Cumolhydroperoxid ≥80%

Lieferant: Roth Carl

ROTH4460.1EA 0 CHF
ROTH4460.1
Cumolhydroperoxid ≥80%
Cumolhydroperoxid
Formel: C₉H₁₂O₂
MW: 152,20 g/mol
MDL Number: MFCD00002129
CAS-Nummer: 80-15-9
EINECS: 201-254-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR