3-Bromphenylhydraziniumchlorid
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
3-Bromophenylhydrazine hydrochloride, 1-(3-Bromophenyl)hydrazine hydrochloride
AS21135G25
AS21135G100
ACTIAS21135G25EA
87
CHF
ACTIAS21135G25
ACTIAS21135G100
3-Bromphenylhydraziniumchlorid
3-Bromphenylhydraziniumchlorid
Formel:
C₆H₈BrClN₂ MW: 223,5 g/mol |
MDL Number:
MFCD00012933 CAS-Nummer: 27246-81-7 EINECS: 248-362-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...