4-Amino-2-methylpyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Amino-2-picoline, 2-Methyl-4-pyridylamine, 2-Methylpyridin-4-amine

AS1631G5 AS1631G100
ACTIAS1631G5EA 0 CHF
ACTIAS1631G5 ACTIAS1631G100
4-Amino-2-methylpyridin
4-Amino-2-methylpyridin
Formel: C₆H₈N₂
MW: 108.14 g/mol
MDL Number: MFCD00186509
CAS-Nummer: 18437-58-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR