5-Amino-2,3-dichlorpyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2,3-Dichloropyridin-5-amine, 2,3-Dichloro-5-pyridylamine, 3-Amino-5,6-dichloropyridine

AS7230G5 AS7230G1 AS7230G25
ACTIAS7230G5EA 56 CHF
ACTIAS7230G5 ACTIAS7230G1 ACTIAS7230G25
5-Amino-2,3-dichlorpyridin
5-Amino-2,3-dichlorpyridin
Formel: C₅H₄Cl₂N₂
MW: 163,01 g/mol
MDL Number: MFCD03840434
CAS-Nummer: 98121-41-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR