EPPS (HEPPS, 4-(2-Hydroxyethyl)-1-piperazinpropansulfonsäure)
Lieferant: Roth Carl
Achtung
Synonyms: 4-(2-Hydroxyethyl)-1-piperazinepropanesulphonic acid, N-(2-Hydroxyethyl)piperazine-N′-(3-propanesulphonic acid), HEPPS, 3-[4-(2-Hydroxyethyl)-1-piperazinyl]propanesulphonic acid, 3-[4-(2-Hydroxyethyl)-1-piperazinyl]-propanesulphonic acid, 4-(2-Hydroxyethyl)piperazine-1-propanesulphonic acid
9164.1 9164.2
ROTH9164.1EA 31.8 CHF
ROTH9164.1 ROTH9164.2
EPPS (HEPPS, 4-(2-Hydroxyethyl)-1-piperazinpropansulfonsäure)
EPPS (HEPPS, 4-(2-Hydroxyethyl)-1-piperazinpropansulfonsäure)
Formel: C₉H₂₀N₂O₄S
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00006160
CAS-Nummer: 16052-06-5
EINECS: 240-198-8