Actinomycin D

Lieferant: EUROCLONE

EURLEMR082025EA 0 CHF
EURLEMR082025 EURLEMR082005 EURLEMR082100
Actinomycin D
Actinomycin D
Formel: C₆₂H₈₆N₁₂O₁₆
MW: 1255,5 g/mol
MDL Number: MFCD00005033
CAS-Nummer: 50-76-0
UN: 3462
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR