Deoxythymidine triphosphate (dTTP)

Lieferant: EUROCLONE

EMR073025 EMR073100
EURLEMR073025EA 0 CHF
EURLEMR073025 EURLEMR073100
Deoxythymidine triphosphate (dTTP)
Deoxythymidine triphosphate (dTTP)
Formel: C₁₀H₁₇N₂O₁₃P₃
MDL Number: MFCD00080338
CAS-Nummer: 18423-43-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR