Inosin

Lieferant: EUROCLONE

EMR156010 EMR156025 EMR156100
EURLEMR156010EA 0 CHF
EURLEMR156010 EURLEMR156025 EURLEMR156100
Inosin
Inosin
Formel: C₁₀H₁₂N₄O₅
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00066770
CAS-Nummer: 58-63-9
EINECS: 200-390-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR