Inosin

Lieferant: EHRENSTORFER GMBH

EHERC14328000EA 109 CHF
EHERC14328000
Inosin
Inosin
Formel: C₁₀H₁₂N₄O₅
MW: 268,23 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00066770
CAS-Nummer: 58-63-9
EINECS: 200-390-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR