Inosin, Millipore®
Lieferant: Merck Millipore Calbiochem
4060-50EA
346
CHF
4060-50
Inosin, Millipore®
Inosin
Formel:
C₁₀H₁₂N₄O₅ MW: 268,23 g/mol Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00066770 CAS-Nummer: 58-63-9 EINECS: 200-390-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...