Puromycin Dihydrochlorid

Lieferant: EUROCLONE
Achtung

Synonyms: (S)-2-Amino-N-((2S,3S,4R,5R)-5-(6-(Dimethylamino)-9H-purin-9-yl)-4-hydroxy-2-(hydroxymethyl)tetrahydrofuran-3-yl)-3-(4-methoxyphenyl)propanamide dihydrochloride

EMR317005 EMR317010 EMR317025 EMR317100
EURLEMR317005EA 0 CHF
EURLEMR317005 EURLEMR317010 EURLEMR317025 EURLEMR317100
Puromycin Dihydrochlorid
Puromycin Dihydrochlorid
Formel: C₂₂H₂₉N₇O₅·2HCl
Lager Temperatur: Gefrierschrank
MDL Number: MFCD00012691
CAS-Nummer: 58-58-2
EINECS: 200-387-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR