Ponceau S

Lieferant: EUROCLONE
Achtung

Synonyms: 3-Hydroxy-4-(2-sulpho-4-[4-sulphophenylazo]phenylazo)-2,7-naphthalenedisulphonic acid sodium salt, CI 27195, Ponceau, 3-Hydroxy-4-((2-sulpho-4-((4-sulphophenyl)diazenyl)phenyl)diazenyl)naphthalene-2,7-disulphonic acid, tetrasodium salt

EMR195010 EMR195100
EURLEMR195010EA 0 CHF
EURLEMR195010 EURLEMR195100
Ponceau S
Ponceau S
Formel: C₂₂H₁₂N₄Na₄O₁₃S₄
MW: 760.56 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00003892
CAS-Nummer: 6226-79-5
EINECS: 228-319-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR