Creatinphosphat Dinatriumsalz Tetrahydrat
Lieferant: Molekula
MOLE14577248-5GEA
0
CHF
MOLE14577248-5G
Creatinphosphat Dinatriumsalz Tetrahydrat
Creatinphosphat Dinatriumsalz Tetrahydrat
Formel:
C₄H₈N₃Na₂O₅P·4H₂O MW: 327,14 g/mol (255,07 anhy) Lager Temperatur: Gefrierschrank |
MDL Number:
MFCD00150192 CAS-Nummer: 71519-72-7 EINECS: 213-074-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...