DEAE-Dextran

Lieferant: EUROCLONE

EMR120025 EMR120100
EURLEMR120025EA 0 CHF
EURLEMR120025 EURLEMR120100
DEAE-Dextran
DEAE-Dextran
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00081561
CAS-Nummer: 9015-73-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR