5-Bromuridin
Lieferant: Molekula
Synonyms:
5-Bromo-1-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione
MOLE22067520-1GEA
0
CHF
MOLE22067520-1G
5-Bromuridin
5-Bromuridin
Formel:
C₉H₁₁BrN₂O₆ MW: 323,10 g/mol Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00006528 CAS-Nummer: 957-75-5 EINECS: 213-486-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...