Tetracyclin Hydrochlorid
Lieferant: EUROCLONE
|
Achtung
|
EMR230025
EMR230050
EMR230100
EURLEMR230025EA
0
CHF
EURLEMR230025
EURLEMR230050
EURLEMR230100
Tetracyclin Hydrochlorid
Tetracyclin Hydrochlorid
Formel:
C₂₂H₂₅ClN₂O₈ MW: 480.9 g/mol Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00078142 CAS-Nummer: 64-75-5 EINECS: 200-593-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...