Tetracyclin

Lieferant: EUROCLONE

EMR229025 EMR229100 EMR229250
EURLEMR229025EA 0 CHF
EURLEMR229025 EURLEMR229100 EURLEMR229250
Tetracyclin
Tetracyclin
Formel: C₂₂H₂₄N₂O₈
MW: 444.43 g/mol
MDL Number: MFCD00151232
CAS-Nummer: 60-54-8
EINECS: 200-481-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR