Dextransulfat Natriumsalz

Lieferant: EUROCLONE

EURLEMR124100EA 0 CHF
EURLEMR124100 EURLEMR124050 EURLEMR124250 EURLEMR124025
Dextransulfat Natriumsalz
Dextran sulphate sodium salt
Formel: (C₆H₇Na₃O₁₄S₃)n
MDL Number: MFCD00081551
CAS-Nummer: 9011-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR