Malondialdehyd

Lieferant: Chemicon

CHMCAB5524EA 0 CHF
CHMCAB5524
Malondialdehyd
Malondialdehyd
Formel: C₃H₄O₂
CAS-Nummer: 542-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR