4-Phenylazophenol ≥98%
Lieferant: Thermo Scientific
Achtung
Synonyms: 4-(Phenyldiazenyl)phenol
B25198.06 B25198.14 B25198.22
B25198.06EA 46.7 CHF
B25198.06 B25198.14 B25198.22
4-Phenylazophenol ≥98%
4-Phenylazophenol
Purity: 98% CAS No.: 1689-82-3 Molecular Formula: C12H10N2O Formula Weight: 198.23 Melting Point: 152-155° MDL No.: MFCD00002330
Formel: C₁₂H₁₀N₂O
MW: 198,23 g/mol
Melting Pt: 152…155 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00002330
CAS-Nummer: 1689-82-3
EINECS: 216-880-6