3-Nitro-L-tyrosin

Lieferant: Alpha Diagnostics International

ALPDNITT13-SEA 1100 CHF
ALPDNITT13-S
3-Nitro-L-tyrosin
3-Nitro-L-tyrosin
Formel: C₉H₁₀N₂O₅
MW: 226,3 g/mol
MDL Number: MFCD00007123
CAS-Nummer: 621-44-3
EINECS: 210-688-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR