3-Oxetanon

Lieferant: ACTIVATE SCIENTIFIC

AS6000G1 AS6000G5 AS6000G25
ACTIAS6000G1EA 0 CHF
ACTIAS6000G1 ACTIAS6000G5 ACTIAS6000G25
3-Oxetanon
3-Oxetanon
Formel: C₃H₄O₂
MDL Number: MFCD09263255
CAS-Nummer: 6704-31-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR