9-Fluorenylmethylchlorformiat (Fmoc-Chlorid)

Lieferant: BACHEM BIOCHEMICA

BACE4004776.0025EA 89.9 CHF
BACE4004776.0025
9-Fluorenylmethylchlorformiat (Fmoc-Chlorid)
9-Fluorenylmethylchlorformiat (Fmoc-Chlorid)
Formel: C₁₅H₁₁ClO₂
MW: 258,7 g/mol
MDL Number: MFCD00001138
CAS-Nummer: 28920-43-6
EINECS: 249-313-6
UN: 2923
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR