(R)-(-)-Thiazolidin-4-carbonsäure

Lieferant: Molekula

MOLE24896910-25GEA 0 CHF
MOLE24896910-25G
(R)-(-)-Thiazolidin-4-carbonsäure
(R)-(-)-Thiazolidin-4-carbonsäure
Formel: C₄H₇NO₂S
MW: 133,17 g/mol
MDL Number: MFCD00005212
CAS-Nummer: 34592-47-7
EINECS: 252-106-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR