Denatoniumbenzoat

Lieferant: EHRENSTORFER GMBH

EHERC10661000EA 94.7 CHF
EHERC10661000
Denatoniumbenzoat
Denatoniumbenzoat
Formel: C₂₈H₃₄N₂O₃
MW: 446,59 g/mol
MDL Number: MFCD00031578
CAS-Nummer: 3734-33-6
EINECS: 223-095-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR