L(+)-Ornithin Hydrochlorid
Lieferant: Roth Carl
Synonyms: H-Orn-OH Hydrochloride, H-Orn-OH·HCl
T204.1 T204.2 T204.3
ROTHT204.1EA 21.5 CHF
ROTHT204.1 ROTHT204.2 ROTHT204.3
L(+)-Ornithin Hydrochlorid
L(+)-Ornithin Hydrochlorid
Formel: C₅H₁₂N₂O₂·HCl
MW: 168.62 g/mol
MDL Number: MFCD00064562
CAS-Nummer: 3184-13-2
EINECS: 221-678-6