2-Chlor-4-methylpyrimidin
Lieferant: ACTIVATE SCIENTIFIC
AS5149G5
AS5149G100
ACTIAS5149G5EA
0
CHF
ACTIAS5149G5
ACTIAS5149G100
2-Chlor-4-methylpyrimidin
2-Chloro-4-methylpyrimidine
Formel:
C₅H₅ClN₂ |
MDL Number:
MFCD00054434 CAS-Nummer: 13036-57-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...