(S)-(-)-3-Acetylthio-2-methylpropionsäure
Lieferant: LGC Standards PROMOCHEM
PROOMM0067.04EA
1220
CHF
PROOMM0067.04
(S)-(-)-3-Acetylthio-2-methylpropionsäure
(S)-(-)-3-Acetylthio-2-methylpropionsäure
Formel:
C₆H₁₀O₃S MW: 162,21 g/mol |
CAS-Nummer:
76497-39-7 EINECS: 278-480-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...