Antimon(III)chlorid, GPR RECTAPUR®

Lieferant: VWR Chemicals
Gefahr

Synonyms: Antimontrichlorid

21468.235EA 0 CHF
21468.235
Antimon(III)chlorid, GPR RECTAPUR®
Antimon(III)chlorid
Formel: SbCl₃
MW: 228,12 g/mol
Siedepunkt: 223,5 °C (1013 hPa)
Melting Pt: 73 °C
Dichte: 3,14 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011212
CAS-Nummer: 10025-91-9
EINECS: 233-047-2
UN: 1733
ADR: 8,II
Merck Index: 13,00713

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR