Natriumhydroxymethansulfinat Dihydrat

Lieferant: Roth Carl

ROTH4553.1EA 61.75 CHF
ROTH4553.1
Natriumhydroxymethansulfinat Dihydrat
Natriumhydroxymethansulfinat Dihydrat
Formel: CH₃NaO₃S·2H₂O
MW: 154,12 g/mol
MDL Number: MFCD00150598
CAS-Nummer: 6035-47-8
EINECS: 205-739-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR