Aminoguanidiniumhydrogencarbonat

Lieferant: Molekula

MOLE19361491-500GEA 0 CHF
MOLE19361491-500G
Aminoguanidiniumhydrogencarbonat
Aminoguanidinium hydrogen carbonate
Formel: C₂H₈N₄O₃
MW: 136,11 g/mol
MDL Number: MFCD00012949
CAS-Nummer: 2582-30-1
EINECS: 219-956-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR