1,3-Dichlor-5,5-dimethylhydantoin

Lieferant: Molekula

MOLE16229036-500GEA 0 CHF
MOLE16229036-500G
1,3-Dichlor-5,5-dimethylhydantoin
1,3-Dichlor-5,5-dimethylhydantoin
Formel: C₅H₆Cl₂N₂O₂
MW: 197,02 g/mol
MDL Number: MFCD00003190
CAS-Nummer: 118-52-5
EINECS: 204-258-7
UN: 1479
ADR: 5.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR