4'-Ethoxyacetanilid

Lieferant: EHRENSTORFER GMBH

EHERC16003000EA 72.3 CHF
EHERC16003000
4'-Ethoxyacetanilid
4'-Ethoxyacetanilid
Formel: C₁₀H₁₃NO₂
MW: 179,22 g/mol
MDL Number: MFCD00009094
CAS-Nummer: 62-44-2
EINECS: 200-533-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR