4-Nitrophenylarsonsäure
Lieferant: EHRENSTORFER GMBH
|
Gefahr
|
EHERC15533000EA
235
CHF
EHERC15533000
4-Nitrophenylarsonsäure
4-Nitrophenylarsonsäure
Formel:
C₆H₆AsNO₅ MW: 247,04 g/mol |
MDL Number:
MFCD00024575 CAS-Nummer: 98-72-6 EINECS: 202-695-8 |
Learn more
![](/m-ch.vwr.com/de_CH/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...