1,1'-Dibromferrocen ≥96%

Lieferant: Thermo Scientific
Achtung

L17045.22EA 0 CHF
L17045.22
1,1'-Dibromferrocen ≥96%
1,1'-Dibromferrocen
Formel: C₁₀Br₂Fe
MW: 335,76 g/mol
Melting Pt: 50…51 °C
Lager Temperatur: Kühlschrank
MDL Number: MFCD01075755
CAS-Nummer: 1293-65-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR