Kaliumbromat
Lieferant: Molekula
|
Gefahr
|
MOLE61847586-1KGEA
0
CHF
MOLE61847586-1KG
Kaliumbromat
Potassium bromate
Formel:
KBrO₃ MW: 167 g/mol Melting Pt: 350 °C Dichte: 3,218 g/cm³ (20 °C) Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00011359 CAS-Nummer: 7758-01-2 EINECS: 231-829-8 UN: 1484 ADR: 5.1,II Merck Index: 12,07779 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...