2,7-Dibromfluoren ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A12437.09
A12437.14
A12437.22
AB115292.0025
A12437.09EA
86
CHF
A12437.09
A12437.14
A12437.22
ABCRAB115292.0025
2,7-Dibromfluoren ≥98%
2,7-Dibromfluoren
Purity: 98%
CAS No.: 16433-88-8
Molecular Formula: C13H8Br2
Formula Weight: 324.03
Melting Point: 164-167°
Beilstein Reference: 2049205
Formel:
C₁₃H₈Br₂ Melting Pt: 164…167 °C Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00019048 CAS-Nummer: 16433-88-8 |