2-Dodecylcyclobutanon, Supelco®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

44197-5MGEA 218 CHF
44197-5MG
2-Dodecylcyclobutanon, Supelco®
2-Dodecylcyclobutanone
Formel: C₁₆H₃₀O
MW: 238,41 g/mol
Siedepunkt: 322 °C (1013 hPa)
Melting Pt: 28…31 °C
Lager Temperatur: Raumtemperatur
CAS-Nummer: 35493-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR