TES (2-(Tris(hydroxymethyl)methylamino)ethansulfonsäure)
Lieferant: Molekula
|
Achtung
|
MOLE17596980-100GEA
0
CHF
MOLE17596980-100G
TES (2-(Tris(hydroxymethyl)methylamino)ethansulfonsäure)
TES (2-(Tris(hydroxymethyl)methylamino)ethansulfonsäure)
Formel:
C₆H₁₅NO₆S MW: 229,25 g/mol Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00007532 CAS-Nummer: 7365-44-8 EINECS: 230-906-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...