Brij® 58 10% in wässriger Lösung

Lieferant: EUROCLONE

Synonyms: Polyoxyethylene (20) cetyl ether, Polyethylene glycol hexadecyl ether

EMR104025 EMR104050 EMR104100
EURLEMR104025EA 0 CHF
EURLEMR104025 EURLEMR104050 EURLEMR104100
Brij® 58 10% in wässriger Lösung
Brij® 58
Formel: C₅₆H₁₁₄O₂₁
MDL Number: MFCD00080892
CAS-Nummer: 9004-95-9
EINECS: 500-014-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR