Brij® 58

Lieferant: EUROCLONE

Synonyms: Polyoxyethylene (20) cetyl ether, Polyethylene glycol hexadecyl ether

EMR103001 EMR103250 EMR103500
EURLEMR103001EA 0 CHF
EURLEMR103001 EURLEMR103250 EURLEMR103500
Brij® 58
Brij® 58
Formel: C₅₆H₁₁₄O₂₁
MDL Number: MFCD00080892
CAS-Nummer: 9004-95-9
EINECS: 500-014-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR