Brij® 35

Lieferant: EUROCLONE
Achtung

EMR101001 EMR101250 EMR101500
EURLEMR101001EA 0 CHF
EURLEMR101001 EURLEMR101250 EURLEMR101500
Brij® 35
Brij® 35
Formel: C₅₈H₁₁₈O₂₄
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00080891
CAS-Nummer: 9002-92-0
EINECS: 500-002-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR