3,5-Di-tert-butylphenol

Lieferant: EHRENSTORFER GMBH

Synonyms: 1,3-Di(dimethylethyl)-5-hydroxybenzol

EHERC12254800EA 367 CHF
EHERC12254800
3,5-Di-tert-butylphenol
3,5-Di-tert-butylphenol
Formel: C₁₄H₂₂O
MW: 206,33 g/mol
Siedepunkt: 277 °C (1013 hPa)
Melting Pt: 87…89 °C
MDL Number: MFCD00008829
CAS-Nummer: 1138-52-9
EINECS: 214-513-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR