Benzyldimethyldodecylammoniumbromid

Lieferant: LGC Standards PROMOCHEM

PROOFPC0141EA 373 CHF
PROOFPC0141
Benzyldimethyldodecylammoniumbromid
Benzyldimethyldodecylammoniumbromid
Formel: C₂₁H₃₈BrN
MW: 384,44 g/mol
MDL Number: MFCD00011768
CAS-Nummer: 7281-04-1
EINECS: 230-698-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR