L(+)-Citrullin

Lieferant: Chemicon

CHMCAB5612EA 1670 CHF
CHMCAB5612
L(+)-Citrullin
L(+)-Citrullin
Formel: C₆H₁₃N₃O₃
MW: 175,19 g/mol
MDL Number: MFCD00064397
CAS-Nummer: 372-75-8
EINECS: 206-759-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR