Gramin

Lieferant: LATOXAN

Synonyms: 1-(1H-Indol-3-yl)-N,N-dimethylmethanamine, 3-(Dimethylaminomethyl)-Indole

L6088-100 L6088-500
LATOL6088-100EA 0 CHF
LATOL6088-100 LATOL6088-500
Gramin
Gramin
Formel: C₁₁H₁₄N₂
MW: 174.25 g/mol
MDL Number: MFCD00005629
CAS-Nummer: 87-52-5
EINECS: 201-749-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR