o-Dianisidin, Supelco®

Lieferant: Merck

Synonyms: 3,3'-Dimethoxybenzidin

M-Clarity™ Qualitätsstufe = MQ100

33430-50GEA 0 CHF
33430-50G 33430-10G
o-Dianisidin, Supelco®
o-Dianisidin
Formel: C₁₄H₁₆N₂O₂
MW: 244,29 g/mol
MDL Number: MFCD00008372
CAS-Nummer: 119-90-4
EINECS: 204-355-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR