Erythrosin B Dinatriumsalz

Lieferant: Roth Carl
Achtung

Synonyms: Erythrosin 239C.I. 45430

ROTH0331.1EA 53.2 CHF
ROTH0331.1
Erythrosin B Dinatriumsalz
Erythrosin B Dinatriumsalz
Formel: C₂₀H₆I₄Na₂O₅
MW: 879,86 g/mol
MDL Number: MFCD00144257
CAS-Nummer: 16423-68-0
EINECS: 240-474-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR