Serotonin

Lieferant: Chemicon

CHMCMAB352EA 941 CHF
CHMCMAB352
Serotonin
Serotonin
Formel: C₁₀H₁₂N₂O
CAS-Nummer: 50-67-9
EINECS: 200-058-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR