4-Fluoranisol

Lieferant: Molekula

MOLE54634510-100GEA 0 CHF
MOLE54634510-100G
4-Fluoranisol
4-Fluoranisol
Formel: C₇H₇FO
MW: 126,13 g/mol
Siedepunkt: 157 °C (1013 hPa)
Melting Pt: –43,5 °C
Dichte: 1,1178 g/cm³ (18.3 °C)
MDL Number: MFCD00000348
CAS-Nummer: 459-60-9
EINECS: 207-295-7
UN: 3271
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR